Gene Transcripts

Gene Symbol
Elovl7
Entrez Gene ID
361895
Gene Name
ELOVL fatty acid elongase 7
Gene Aliases
Chromosome Location
Chr. 2: 40373211 - 40386643 on Build Rnor_6.0
UniGene ID
Rn.105650
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191844.1 NP_001178773.1 288

View Full Product Details