Gene Transcripts

Gene Symbol
Coq10a
Entrez Gene ID
362810
Gene Name
coenzyme Q10 homolog A (S. cerevisiae)
Gene Aliases
RGD1562447
Chromosome Location
Chr. 7: 2780930 - 2788190 on Build Rnor_6.0
UniGene ID
Rn.25373
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108727.1 NP_001102197.1 3 408
XM_006240759.2 XP_006240821.1 3 1577

View Full Product Details