Gene Transcripts

Gene Symbol
Fzd8
Entrez Gene ID
364754
Gene Name
frizzled class receptor 8
Gene Aliases
Chromosome Location
Chr. 17: 62259905 - 62267239 on Build Rnor_6.0
UniGene ID
Rn.234505
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044251.2 NP_001037716.1 1 1539
XM_006254142.2 XP_006254204.1 2 2967
GenBank mRNA BC100088.1 - 1 1542

Gene Symbol
LOC100909849
Entrez Gene ID
100909849
Gene Name
frizzled-8-like
Gene Aliases
Chromosome Location
Chr. 17: 62997442 - 63002835 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003751721.3 XP_003751769.1 2967

View Full Product Details