Gene Transcripts

Gene Symbol
Sult1a1
Entrez Gene ID
83783
Gene Name
sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1
Gene Aliases
ASTIV, Mx-ST, PST-1, St1a1, Stm, Stp1, Sult1a3
Chromosome Location
Chr. 1: 198100586 - 198104106 on Build Rnor_6.0
UniGene ID
Rn.1507
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031834.1 NP_114022.1 3, 4 338
GenBank mRNA AF394783.1 - 2, 3 261
FQ209630.1 - 3, 4 347
FQ210886.1 - 3, 4 343
FQ214244.1 - 3, 4 343
FQ218459.1 - 3, 4 343
FQ218506.1 - 3, 4 352
FQ218658.1 - 3, 4 342
FQ219178.1 - 3, 4 343
FQ219521.1 - 3, 4 343
FQ219948.1 - 3, 4 343
L19998.1 - 3, 4 338
X52883.1 - 3, 4 326
X68640.1 - 2, 3 243

View Full Product Details