Gene Transcripts

Gene Symbol
Gosr1
Entrez Gene ID
94189
Gene Name
golgi SNAP receptor complex member 1
Gene Aliases
GOS28, p28
Chromosome Location
Chr. 10: 63405528 - 63442257 on Build Rnor_6.0
UniGene ID
Rn.8228
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053584.2 NP_446036.1 2 83
XM_006246914.1 XP_006246976.1 2 157
GenBank mRNA BC126068.1 - 2 87

View Full Product Details