Gene Transcripts

Gene Symbol
Hic2
Entrez Gene ID
287940
Gene Name
hypermethylated in cancer 2
Gene Aliases
Chromosome Location
Chr. 11: 87988213 - 88016825 on Build Rnor_6.0
UniGene ID
Rn.219216
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105862.1 NP_001099332.1 3 1759

View Full Product Details