Gene Transcripts

Gene Symbol
Spag7
Entrez Gene ID
303260
Gene Name
sperm associated antigen 7
Gene Aliases
Chromosome Location
Chr. 10: 57284989 - 57291146 on Build Rnor_6.0
UniGene ID
Rn.219156
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107016.1 NP_001100486.1 6 635
GenBank mRNA FQ233279.1 - 6 507

View Full Product Details