Gene Transcripts

Gene Symbol
Stx5
Entrez Gene ID
65134
Gene Name
syntaxin 5
Gene Aliases
Stx5a
Chromosome Location
Chr. 1: 224939497 - 224955658 on Build Rnor_6.0
UniGene ID
Rn.5782
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031704.2 NP_113892.2 9 877
XM_006230997.2 XP_006231059.1 8 1420
GenBank mRNA BC085724.1 - 8 1203
BC127489.1 - 9 877 7376571
L20822.1 - 8 707

View Full Product Details