Gene Transcripts

Gene Symbol
Cfd
Entrez Gene ID
54249
Gene Name
complement factor D (adipsin)
Gene Aliases
Adn, Df, EVE
Chromosome Location
Chr. 7: 12634216 - 12635939 on Build Rnor_6.0
UniGene ID
Rn.16172
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077642.1 NP_001071110.1 2 122
GenBank mRNA M92059.1 - 2 119
S73894.1 - 2 122

View Full Product Details