Gene Transcripts

Gene Symbol
Evx2
Entrez Gene ID
288155
Gene Name
even-skipped homeobox 2
Gene Aliases
Chromosome Location
Chr. 3: 61564917 - 61581598 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_221512.8 XP_221512.5 1 207

View Full Product Details