Gene Transcripts

Gene Symbol
Hoxb1
Entrez Gene ID
303491
Gene Name
homeo box B1
Gene Aliases
Chromosome Location
Chr. 10: 84214358 - 84215812 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_220896.6 XP_220896.4 2 605

View Full Product Details