Gene Transcripts

Gene Symbol
Dnmbp
Entrez Gene ID
309362
Gene Name
dynamin binding protein
Gene Aliases
Chromosome Location
Chr. 1: 263625503 - 263718782 on Build Rnor_6.0
UniGene ID
Rn.11811
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006231570.2 XP_006231632.1 10 3125
XM_008760486.1 XP_008758708.1 10 3634

View Full Product Details