Gene Transcripts

Gene Symbol
Egfem1
Entrez Gene ID
310269
Gene Name
EGF-like and EMI domain containing 1
Gene Aliases
RGD1307225
Chromosome Location
Chr. 2: 116526140 - 117152634 on Build Rnor_6.0
UniGene ID
Rn.218693
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107663.1 NP_001101133.1 11 1549
XM_008760923.1 XP_008759145.1 11 1637

View Full Product Details