Gene Transcripts

Gene Symbol
Btaf1
Entrez Gene ID
368042
Gene Name
BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, 170kDa
Gene Aliases
RGD1564130
Chromosome Location
Chr. 1: 255564199 - 255654932 on Build Rnor_6.0
UniGene ID
Rn.1237
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191917.1 NP_001178846.1 20 2506
XM_006231332.2 XP_006231394.1 20 2792
XM_006231334.1 XP_006231396.1 17 2168

View Full Product Details