Gene Transcripts

Gene Symbol
Adm2
Entrez Gene ID
399475
Gene Name
adrenomedullin 2
Gene Aliases
Imd
Chromosome Location
Chr. 7: 130296895 - 130298641 on Build Rnor_6.0
UniGene ID
Rn.145378
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201426.1 NP_958829.1 3 392
GenBank mRNA AB121036.1 - 3 392
AB181297.1 - 3 401
AY590103.1 - 2 220

View Full Product Details