Gene Transcripts

Gene Symbol
Krt77
Entrez Gene ID
406226
Gene Name
keratin 77, type II
Gene Aliases
Kb39
Chromosome Location
Chr. 7: 143461423 - 143473840 on Build Rnor_6.0
UniGene ID
Rn.129725
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008807.1 NP_001008807.1 1 388
XM_006242435.2 XP_006242497.1 1 531

View Full Product Details