Gene Transcripts

Gene Symbol
Clec4a
Entrez Gene ID
474143
Gene Name
C-type lectin domain family 4, member A
Gene Aliases
Clecsf6, Dcir1
Chromosome Location
Chr. 4: 156050040 - 156067518 on Build Rnor_6.0
UniGene ID
Rn.24032
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005899.2 NP_001005899.2 4 373
GenBank mRNA AY363176.1 - 4 419
AY494060.1 - 4 372
AY581051.1 - 4 373

View Full Product Details