Gene Transcripts

Gene Symbol
Zfp61
Entrez Gene ID
499094
Gene Name
zinc finger protein 61
Gene Aliases
Chromosome Location
Chr. 1: 81122789 - 81130820 on Build Rnor_6.0
UniGene ID
Rn.122263
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC079015.1 - 6 1284

View Full Product Details