Gene Transcripts

Gene Symbol
Mapk1ip1
Entrez Gene ID
499280
Gene Name
mitogen-activated protein kinase 1 interacting protein 1
Gene Aliases
RGD1565431
Chromosome Location
Chr. 1: 211195440 - 211205778 on Build Rnor_6.0
UniGene ID
Rn.224616
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122782.1 NP_001116254.1 1 1183
XM_006230487.2 XP_006230549.1 3 2327
XM_006230488.2 XP_006230550.1 2 2618
XM_006230489.2 XP_006230551.1 4 1492
XM_006230490.2 XP_006230552.1 3 1369
XM_006230491.2 XP_006230553.1 2 1248
XM_008760006.1 XP_008758228.1 3 1783
XM_008760007.1 XP_008758229.1 2 1660
GenBank mRNA BC158819.1 - 1 1183

View Full Product Details