Gene Transcripts

Gene Symbol
Ssuh2
Entrez Gene ID
500286
Gene Name
ssu-2 homolog
Gene Aliases
RGD1562010
Chromosome Location
Chr. 4: 144301922 - 144322197 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002729443.4 XP_002729489.2 12 1111
XM_006237061.2 XP_006237123.1 12 1108

View Full Product Details