Gene Transcripts

Gene Symbol
Prl2c1
Entrez Gene ID
502122
Gene Name
Prolactin family 2, subfamily c, member 1
Gene Aliases
RGD1562878
Chromosome Location
Chr. 17: 38849932 - 38858391 on Build Rnor_6.0
UniGene ID
Rn.194900
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044271.1 NP_001037736.1 5 670
GenBank mRNA DQ329281.1 - 5 670

View Full Product Details