Gene Transcripts

Gene Symbol
Smn1
Entrez Gene ID
64301
Gene Name
survival of motor neuron 1, telomeric
Gene Aliases
Smn
Chromosome Location
Chr. 2: 30360101 - 30371147 on Build Rnor_6.0
UniGene ID
Rn.1119
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022509.1 NP_071954.1 2, 3 182
GenBank mRNA U75369.1 - 2, 3 182

View Full Product Details