Gene Transcripts

Gene Symbol
Pip5kl1
Entrez Gene ID
687758
Gene Name
phosphatidylinositol-4-phosphate 5-kinase-like 1
Gene Aliases
Chromosome Location
Chr. 3: 11591226 - 11597505 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749451.3 XP_003749499.2 1 1457
XM_006234011.2 XP_006234073.1 1 1457

View Full Product Details