Gene Transcripts

Gene Symbol
Wscd2
Entrez Gene ID
360824
Gene Name
WSC domain containing 2
Gene Aliases
Wscd2-ps1
Chromosome Location
Chr. 12: 48817320 - 48857724 on Build Rnor_6.0
UniGene ID
Rn.178658
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271178.1 NP_001258107.1 3 721

View Full Product Details