Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCND3
Entrez Gene ID
695215
Gene Name
cyclin D3
Gene Aliases
EGK_14897
Chromosome Location
Chr.4: 127327473 - 127453384 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265987.1 NP_001252916.1 - 556 69
XM_028846630.1 XP_028702463.1 - 715 69
GenBank mRNA JU322013.1 - - 556 69
JV048616.1 - - 556 69

View Full Product Details