Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FCRL3
Entrez Gene ID
718801
Gene Name
Fc receptor like 3
Gene Aliases
-
Chromosome Location
Chr.1: 93290912 - 93314478 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015112565.2 XP_014968051.2 - 1851 122

View Full Product Details