Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDK6
Entrez Gene ID
701822
Gene Name
cyclin dependent kinase 6
Gene Aliases
EGK_13940
Chromosome Location
Chr.3: 113285491 - 113523622 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261307.1 NP_001248236.1 - 853 139
XM_015134230.2 XP_014989716.1 - 1513 139
XM_015134231.2 XP_014989717.1 - 997 139
XM_028844866.1 XP_028700699.1 - 1583 139
XM_028844867.1 XP_028700700.1 - 1716 139
GenBank mRNA JU319885.1 - - 719 139
JU319886.1 - - 719 139
JU471047.1 - - 853 139
JU471048.1 - - 853 139

View Full Product Details