Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO42
Entrez Gene ID
698061
Gene Name
F-box protein 42
Gene Aliases
EGK_00284
Chromosome Location
Chr.1: 208431552 - 208529613 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266406.1 NP_001253335.1 - 1066 66
GenBank mRNA JU476075.1 - - 1066 66
JV047426.1 - - 1051 66

View Full Product Details