The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_001111024.4 | XP_001111024.2 | - | 900 | 100 |
XM_028831720.1 | XP_028687553.1 | - | 276 | 100 | |
XM_028831721.1 | XP_028687554.1 | - | 276 | 100 | |
XM_028831722.1 | XP_028687555.1 | - | 181 | 100 | |
XM_028831723.1 | XP_028687556.1 | - | 175 | 100 | |
XM_028831724.1 | XP_028687557.1 | - | 278 | 100 | |
XM_028831725.1 | XP_028687558.1 | - | 854 | 100 | |
XM_028831726.1 | XP_028687559.1 | - | 234 | 100 | |
XM_028831727.1 | XP_028687560.1 | - | 345 | 100 | |
GenBank mRNA | JU323176.1 | - | - | 211 | 100 |
JU478147.1 | - | - | 184 | 100 | |
JV678509.1 | - | - | 212 | 100 |