Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAQR3
Entrez Gene ID
697120
Gene Name
progestin and adipoQ receptor family member 3
Gene Aliases
-
Chromosome Location
Chr.5: 54813791 - 54841145 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257693.1 NP_001244622.1 - 951 114
XM_015138441.2 XP_014993927.1 - 998 114
XM_028847985.1 XP_028703818.1 - 998 114
GenBank mRNA JU320618.1 - - 951 114
JU474452.1 - - 833 114
JV044586.1 - - 956 114

View Full Product Details