The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001266275.1 | NP_001253204.1 | - | 223 | 146 |
XM_015140829.2 | XP_014996315.1 | - | 316 | 146 | |
XM_015140830.2 | XP_014996316.1 | - | 316 | 146 | |
GenBank mRNA | JU475994.1 | - | - | 223 | 146 |
JV063291.1 | - | - | 191 | 146 |