Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPS14
Entrez Gene ID
708475
Gene Name
mitochondrial ribosomal protein S14
Gene Aliases
EGK_01989
Chromosome Location
Chr.1: 32277484 - 32286714 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001104412.4 XP_001104412.1 - 210 76
GenBank mRNA JV566399.1 - - 57 76

View Full Product Details