Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FSHB
Entrez Gene ID
697317
Gene Name
follicle stimulating hormone subunit beta
Gene Aliases
EGK_06393
Chromosome Location
Chr.14: 35184319 - 35188575 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001087914.3 XP_001087914.1 - 296 76

View Full Product Details