Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNT10A
Entrez Gene ID
700692
Gene Name
Wnt family member 10A
Gene Aliases
-
Chromosome Location
Chr.12: 106313728 - 106328807 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001095740.4 XP_001095740.1 - 1906 75
GenBank mRNA JV724096.1 - - 53 75

View Full Product Details