Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MCMDC2
Entrez Gene ID
704020
Gene Name
minichromosome maintenance domain containing 2
Gene Aliases
-
Chromosome Location
Chr.8: 67523173 - 67575168 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015145491.2 XP_015000977.2 - 1636 136
XM_028852704.1 XP_028708537.1 - 1585 136
GenBank mRNA JV053109.1 - - 182 136
JV572756.1 - - 177 136

View Full Product Details