Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GBX2
Entrez Gene ID
693901
Gene Name
gastrulation brain homeobox 2
Gene Aliases
-
Chromosome Location
Chr.12: 123586832 - 123589993 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001082998.3 XP_001082998.1 - 949 89

View Full Product Details