Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZG16
Entrez Gene ID
706522
Gene Name
zymogen granule protein 16
Gene Aliases
EGK_12648
Chromosome Location
Chr.20: 27583601 - 27616187 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028840825.1 XP_028696658.1 - 169 115

View Full Product Details