Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KAT6A
Entrez Gene ID
703800
Gene Name
lysine acetyltransferase 6A
Gene Aliases
MYST3
Chromosome Location
Chr.8: 42518176 - 42641485 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015145294.2 XP_015000780.2 - 2328 149
XM_028852629.1 XP_028708462.1 - 3793 149
XM_028852630.1 XP_028708463.1 - 3772 149
GenBank mRNA JU493760.1 - - 3762 149

View Full Product Details