The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_001100732.4 | XP_001100732.2 | - | 3144 | 116 |
XM_015124042.2 | XP_014979528.2 | - | 2865 | 116 | |
XM_028850415.1 | XP_028706248.1 | - | 2990 | 116 | |
GenBank mRNA | JU334889.1 | - | - | 3095 | 116 |
JU334890.1 | - | - | 3095 | 116 | |
JU334891.1 | - | - | 3095 | 116 | |
JU475899.1 | - | - | 3042 | 116 | |
JV047300.1 | - | - | 3101 | 116 | |
JV047301.1 | - | - | 3080 | 116 | |
JV636208.1 | - | - | 3058 | 116 |