Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SUCO
Entrez Gene ID
705776
Gene Name
SUN domain containing ossification factor
Gene Aliases
C1H1orf9
Chromosome Location
Chr.1: 29836904 - 29918532 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001100732.4 XP_001100732.2 - 899 139
XM_015124042.2 XP_014979528.2 - 641 139
XM_028850415.1 XP_028706248.1 - 905 139
GenBank mRNA JU334889.1 - - 850 139
JU334890.1 - - 850 139
JU334891.1 - - 850 139
JU475899.1 - - 797 139
JV047300.1 - - 856 139
JV047301.1 - - 856 139
JV636208.1 - - 813 139

View Full Product Details