Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HINT3
Entrez Gene ID
712779
Gene Name
histidine triad nucleotide binding protein 3
Gene Aliases
-
Chromosome Location
Chr.4: 29478820 - 29501621 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194369.3 NP_001181298.1 - 510 145
GenBank mRNA JU323281.1 - - 569 145
JU470952.1 - - 424 145
JV045448.1 - - 463 145
JV636750.1 - - 543 145
JV669625.1 - - 510 145

View Full Product Details