Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC50A1
Entrez Gene ID
717424
Gene Name
solute carrier family 50 member 1
Gene Aliases
EGK_01377, RAG1AP1
Chromosome Location
Chr.1: 95890792 - 95894247 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001115177.4 XP_001115177.1 - 177 62
XM_001115244.4 XP_001115244.1 - 205 62
XM_015111163.2 XP_014966649.1 - 42 62
XM_028828400.1 XP_028684233.1 - 177 62
GenBank mRNA JU321242.1 - - 168 62
JU322404.1 - - 168 62
JU473587.1 - - 188 62
JU473588.1 - - 188 62
JV047654.1 - - 186 62
JV047655.1 - - 186 62

View Full Product Details