Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM255A
Entrez Gene ID
696050
Gene Name
transmembrane protein 255A
Gene Aliases
EGK_20885, FAM70A
Chromosome Location
Chr.X: 116471498 - 116523677 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193758.1 NP_001180687.1 - 306 65
XM_001085757.2 XP_001085757.1 - 306 65
XM_028841669.1 XP_028697502.1 - 306 65
GenBank mRNA JU335206.1 - - 198 65
JU335207.1 - - 198 65
JV047488.1 - - 124 65
JV636245.1 - - 273 65

View Full Product Details