Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NKG2D
Entrez Gene ID
574240
Gene Name
NKG2D protein
Gene Aliases
KLRK1, NKG2-D
Chromosome Location
Chr.11: 9359059 - 9376449 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032889.1 NP_001028061.1 - 443 113
XM_015150990.2 XP_015006476.2 - 618 113
GenBank mRNA AF190943.1 - - 443 113
AF190944.1 - - 350 113
AJ554302.1 - - 430 113
JU474751.1 - - 571 113

View Full Product Details