Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPOPL
Entrez Gene ID
704203
Gene Name
speckle type BTB/POZ protein like
Gene Aliases
EGK_04395
Chromosome Location
Chr.12: 25579528 - 25644728 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266382.1 NP_001253311.1 - 943 70
XM_015110018.2 XP_014965504.2 - 1214 70
XM_015110019.2 XP_014965505.2 - 1144 70
XM_015110020.2 XP_014965506.1 - 1043 70
GenBank mRNA JU334320.1 - - 1051 70
JU475422.1 - - 943 70

View Full Product Details