Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RRAS2
Entrez Gene ID
699700
Gene Name
RAS related 2
Gene Aliases
-
Chromosome Location
Chr.14: 51265645 - 51344385 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260560.1 NP_001247489.1 - 237 74
GenBank mRNA JU327851.1 - - 165 74
JV044968.1 - - 237 74
JV635708.1 - - 488 74

View Full Product Details