Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MCHR1
Entrez Gene ID
574232
Gene Name
melanin concentrating hormone receptor 1
Gene Aliases
GPR24, MCH1R
Chromosome Location
Chr.10: 10851668 - 10854314 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032885.1 NP_001028057.1 - 189 84
GenBank mRNA AF513988.1 - - 189 84
AY078245.1 - - 83 84
JV579141.1 - - 200 84

View Full Product Details