Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD1D
Entrez Gene ID
613268
Gene Name
CD1d molecule
Gene Aliases
-
Chromosome Location
Chr.1: 92810974 - 92818536 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033114.2 NP_001028286.2 - 101 85
XM_015109844.2 XP_014965330.2 - 477 85
XM_015109854.2 XP_014965340.2 - 477 85
GenBank mRNA AB222829.1 - - 66 85
JU496085.1 - - 101 85

View Full Product Details