Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LEP
Entrez Gene ID
698728
Gene Name
leptin
Gene Aliases
OB
Chromosome Location
Chr.3: 154708773 - 154722828 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042755.1 NP_001036220.1 - 198 88
GenBank mRNA U58492.1 - - 198 88

View Full Product Details