Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NEFL
Entrez Gene ID
641444
Gene Name
neurofilament light
Gene Aliases
EGK_21229
Chromosome Location
Chr.8: 25216264 - 25222081 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265845.2 NP_001252774.1 - 1147 84
GenBank mRNA AY680477.1 - - 279 84
JV528032.1 - - 1097 84
JV600285.1 - - 1147 84

View Full Product Details